Product Details

SNP ID
rs2076598
Assay Type
Functionally tested
NCBI dbSNP Submissions
65
Location
Chr.1:17069026 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GGAGAACTAAGCGAAGGAGGCAAAC[A/G]CCAGGGCCCCTGGCAGGTCAGGGCA
Phenotype
MIM: 607935
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PADI2 PubMed Links

Gene Details

Gene
PADI2
Gene Name
peptidyl arginine deiminase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007365.2 1272 UTR 3 NP_031391.2
XM_017000148.1 1272 UTR 3 XP_016855637.1

View Full Product Details