Product Details

SNP ID
rs10157387
Assay Type
Functionally tested
NCBI dbSNP Submissions
61
Location
Chr.1:17068128 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGAAGGCCTGAATCTGGCTGCTGG[C/T]GAACAAGTTCTTGTCTAGCTGCCTG
Phenotype
MIM: 607935
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PADI2 PubMed Links

Gene Details

Gene
PADI2
Gene Name
peptidyl arginine deiminase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007365.2 2170 UTR 3 NP_031391.2
XM_017000148.1 2170 UTR 3 XP_016855637.1

View Full Product Details