Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001258357.1 | Intron | NP_001245286.1 | |||
NM_001258358.1 | Intron | NP_001245287.1 | |||
NM_001258359.1 | Intron | NP_001245288.1 | |||
NM_002149.3 | Intron | NP_002140.2 | |||
NM_134421.2 | Intron | NP_602293.1 | |||
XM_005246160.1 | Intron | XP_005246217.1 | |||
XM_005246161.1 | Intron | XP_005246218.1 | |||
XM_005246162.1 | Intron | XP_005246219.1 | |||
XM_005246163.1 | Intron | XP_005246220.1 | |||
XM_011510346.2 | Intron | XP_011508648.1 | |||
XM_011510347.1 | Intron | XP_011508649.1 | |||
XM_011510348.1 | Intron | XP_011508650.1 | |||
XM_011510349.1 | Intron | XP_011508651.1 | |||
XM_017003950.1 | Intron | XP_016859439.1 | |||
XM_017003951.1 | Intron | XP_016859440.1 | |||
XM_017003952.1 | Intron | XP_016859441.1 | |||
XM_017003953.1 | Intron | XP_016859442.1 |