Product Details

SNP ID
rs132859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:45167528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGCCAATGATTTGTTTTTGAAAG[G/T]TTATAAGTTCCCAGAATACTAACAC
Phenotype
MIM: 604646
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NUP50 PubMed Links

Gene Details

Gene
NUP50
Gene Name
nucleoporin 50
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007172.3 Intron NP_009103.2
NM_153645.2 Intron NP_705931.1
XM_005261312.1 Intron XP_005261369.1
XM_005261314.1 Intron XP_005261371.1
XM_006724103.1 Intron XP_006724166.1
XM_006724104.1 Intron XP_006724167.1
XM_011529833.1 Intron XP_011528135.1
XM_017028537.1 Intron XP_016884026.1
XM_017028538.1 Intron XP_016884027.1
Gene
NUP50-AS1
Gene Name
NUP50 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details