Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_007172.3 | Intron | NP_009103.2 | |||
NM_153645.2 | Intron | NP_705931.1 | |||
XM_005261312.1 | Intron | XP_005261369.1 | |||
XM_005261314.1 | Intron | XP_005261371.1 | |||
XM_006724103.1 | Intron | XP_006724166.1 | |||
XM_006724104.1 | Intron | XP_006724167.1 | |||
XM_011529833.1 | Intron | XP_011528135.1 | |||
XM_017028537.1 | Intron | XP_016884026.1 | |||
XM_017028538.1 | Intron | XP_016884027.1 |