Product Details
- SNP ID
-
rs104894920
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:83508928 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTGGAACAGTTCGCCAAACAATTC[A/T]AACAAAGAAGAATCAAGTTGGGCTT
- Phenotype
-
MIM: 300039
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
POU3F4
PubMed Links
Gene Details
- Gene
- POU3F4
- Gene Name
- POU class 3 homeobox 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000307.4 |
668 |
Nonsense Mutation |
AAA,TAA |
K202* |
NP_000298.3 |
View Full Product Details