Product Details

SNP ID
rs3888816
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:180928 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGGGCCCCGGCGGGGTGGGTGC[C/G]GGGGGCGTGCGGGGCGGGTGCAGGC
Phenotype
MIM: 141800 MIM: 141850 MIM: 142240 MIM: 607782
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HBA1 PubMed Links

Gene Details

Gene
HBA1
Gene Name
hemoglobin subunit alpha 1
There are no transcripts associated with this gene.

Gene
HBA2
Gene Name
hemoglobin subunit alpha 2
There are no transcripts associated with this gene.

Gene
HBQ1
Gene Name
hemoglobin subunit theta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005331.4 Intron NP_005322.1
Gene
LUC7L
Gene Name
LUC7 like
There are no transcripts associated with this gene.

View Full Product Details