Product Details

SNP ID
rs2008130
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:35087386 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGACTGCCGTAGGATATAGACTGTA[A/C]TGACTGTCTTAATTTTCTTTTTTTT
Phenotype
MIM: 615902
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM177A1 PubMed Links

Gene Details

Gene
FAM177A1
Gene Name
family with sequence similarity 177 member A1
There are no transcripts associated with this gene.

Gene
LOC101927178
Gene Name
uncharacterized LOC101927178
There are no transcripts associated with this gene.

Gene
PPP2R3C
Gene Name
protein phosphatase 2 regulatory subunit B''gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305155.1 Intron NP_001292084.1
NM_001305156.1 Intron NP_001292085.1
NM_017917.3 Intron NP_060387.2
XM_005267782.3 Intron XP_005267839.1
XM_017021388.1 Intron XP_016876877.1
XM_017021389.1 Intron XP_016876878.1

View Full Product Details