Product Details

SNP ID
rs739041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37228959 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGCTGGCAAGAGCAGGGTGCCCTG[A/G]GCTCAGCCCGGAGTCTGGTCACCAC
Phenotype
MIM: 602049 MIM: 182453
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAC2 PubMed Links

Gene Details

Gene
RAC2
Gene Name
ras-related C3 botulinum toxin substrate 2 (rho family, small GTP binding protein Rac2)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002872.4 Intron NP_002863.1
XM_006724286.3 Intron XP_006724349.1
Gene
SSTR3
Gene Name
somatostatin receptor 3
There are no transcripts associated with this gene.

View Full Product Details