Product Details

SNP ID
rs375437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:10713964 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCAGATCTCTTTCCTAGATCAGAA[A/G]AAACAGTATCTGACCACAGAGTTTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADPRM PubMed Links

Gene Details

Gene
ADPRM
Gene Name
ADP-ribose/CDP-alcohol diphosphatase, manganese dependent
There are no transcripts associated with this gene.

Gene
MAGOH2P
Gene Name
mago homolog 2, pseudogene
There are no transcripts associated with this gene.

Gene
TMEM220
Gene Name
transmembrane protein 220
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004313.1 2450 UTR 3 NP_001004313.1
XM_005256634.4 2450 Intron XP_005256691.1
XM_011523835.2 2450 Intron XP_011522137.1
XM_011523836.2 2450 Intron XP_011522138.1
XM_017024616.1 2450 Intron XP_016880105.1
XM_017024617.1 2450 Intron XP_016880106.1

View Full Product Details