Product Details

SNP ID
rs391838
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61788733 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCTCTCCCATGGCCACACCTTC[C/T]CCTCCTGCGCACCCGCTGCTGCCCA
Phenotype
MIM: 600393 MIM: 608329
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FEN1 PubMed Links

Gene Details

Gene
FEN1
Gene Name
flap structure-specific endonuclease 1
There are no transcripts associated with this gene.

Gene
MIR611
Gene Name
microRNA 611
There are no transcripts associated with this gene.

Gene
MYRF
Gene Name
myelin regulatory factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127392.2 Intron NP_001120864.1
NM_013279.3 Intron NP_037411.1
XM_005274222.1 Intron XP_005274279.1
XM_005274223.1 Intron XP_005274280.1
XM_005274224.1 Intron XP_005274281.1
XM_005274225.1 Intron XP_005274282.1
XM_005274226.1 Intron XP_005274283.1
XM_005274227.1 Intron XP_005274284.1
XM_005274228.1 Intron XP_005274285.1
XM_011545234.2 Intron XP_011543536.1
Gene
TMEM258
Gene Name
transmembrane protein 258
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014206.3 Intron NP_055021.1

View Full Product Details