Product Details

SNP ID
rs600661
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:131955201 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGATGGGCATCATCTTTATGGTA[C/T]CATTTATGGCTAAAGATAGGTATCA
Phenotype
MIM: 604443 MIM: 616232
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACSL6 PubMed Links

Gene Details

Gene
ACSL6
Gene Name
acyl-CoA synthetase long-chain family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009185.2 Intron NP_001009185.1
NM_001205247.1 Intron NP_001192176.1
NM_001205248.1 Intron NP_001192177.1
NM_001205250.1 Intron NP_001192179.1
NM_001205251.1 Intron NP_001192180.1
NM_015256.3 Intron NP_056071.2
Gene
MEIKIN
Gene Name
meiotic kinetochore factor
There are no transcripts associated with this gene.

View Full Product Details