Product Details

SNP ID
rs759609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:20425810 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCCTCCTCGGGCAGCCCGGGGGG[C/G]CGCGGCGTTGGGTCGCGGGTCCGGG
Phenotype
MIM: 613619
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SCARF2 PubMed Links

Gene Details

Gene
SCARF2
Gene Name
scavenger receptor class F member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153334.6 2271 Silent Mutation CGC,CGG R727R NP_699165.3
NM_182895.4 2271 Silent Mutation CGC,CGG R722R NP_878315.2
XM_017029065.1 2271 Intron XP_016884554.1

View Full Product Details