Product Details
- SNP ID
-
rs178286
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:20985626 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- GGTCCTGACCCCAGGCAGGCTGGTG[G/T]CTGCCTGTGTGGCAGCCCTGGTGCC
- Phenotype
-
MIM: 600574
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
AIFM3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114940264] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AIFM3
- Gene Name
- apoptosis inducing factor, mitochondria associated 3
There are no transcripts associated with this gene.
- Gene
- LZTR1
- Gene Name
- leucine zipper like transcription regulator 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006767.3 |
|
Intron |
|
|
NP_006758.2 |
View Full Product Details