Product Details

SNP ID
rs174358
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:17563390 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AGATTTGGCTATTGTGTGCTGCAAT[C/G]CCAGCCCCCTTCTACAGCCATGACA
Phenotype
MIM: 607576 MIM: 609303
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CECR2 PubMed Links

Gene Details

Gene
CECR2
Gene Name
CECR2, histone acetyl-lysine reader
There are no transcripts associated with this gene.

Gene
SLC25A18
Gene Name
solute carrier family 25 member 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303484.1 892 Intron NP_001290413.1
NM_031481.2 892 Intron NP_113669.1
XM_011546149.2 892 UTR 5 XP_011544451.1
XM_011546150.2 892 UTR 5 XP_011544452.1
XM_011546151.2 892 Intron XP_011544453.1
XM_011546152.1 892 Intron XP_011544454.1
XM_011546153.2 892 UTR 5 XP_011544455.1
XM_011546154.2 892 Intron XP_011544456.1
XM_017028968.1 892 Intron XP_016884457.1
XM_017028969.1 892 UTR 5 XP_016884458.1
XM_017028970.1 892 UTR 5 XP_016884459.1
XM_017028971.1 892 UTR 5 XP_016884460.1
XM_017028972.1 892 UTR 5 XP_016884461.1
XM_017028973.1 892 UTR 5 XP_016884462.1
XM_017028974.1 892 UTR 5 XP_016884463.1
XM_017028975.1 892 UTR 5 XP_016884464.1

View Full Product Details