Product Details
- SNP ID
-
rs140092
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:29726290 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- ATAAGCCTGATAAGGGTGGGGCCTC[C/T]GCCCAAGCCACCTCGAAGTGTGGCC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CABP7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs144127864] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CABP7
- Gene Name
- calcium binding protein 7
- Gene
- ZMAT5
- Gene Name
- zinc finger matrin-type 5
There are no transcripts associated with this gene.
View Full Product Details