Product Details
- SNP ID
-
rs238741
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:114045084 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGGCCCTCGCGCCTCGTGGCCCTG[A/C]GCGAGCAGAAGGCGCTCAAGACGCT
- Phenotype
-
MIM: 109630
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ADRB1
PubMed Links
Gene Details
- Gene
- ADRB1
- Gene Name
- adrenoceptor beta 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000684.2 |
1038 |
Missense Mutation |
AGC,CGC |
S318R |
NP_000675.1 |
View Full Product Details