Product Details

SNP ID
rs539002
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:44815900 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTACTTTTGCACCAACATATCAAC[C/T]AACTTGACCTAATATTTATAGAATA
Phenotype
MIM: 602947
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929770 PubMed Links

Gene Details

Gene
LOC101929770
Gene Name
uncharacterized LOC101929770
There are no transcripts associated with this gene.

Gene
SUPT3H
Gene Name
SPT3 homolog, SAGA and STAGA complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261823.1 Intron NP_001248752.1
NM_003599.3 Intron NP_003590.1
NM_181356.2 Intron NP_852001.1
XM_011514949.2 Intron XP_011513251.1
XM_011514950.1 Intron XP_011513252.1
XM_011514951.2 Intron XP_011513253.1
XM_011514952.2 Intron XP_011513254.1
XM_011514953.2 Intron XP_011513255.1
XM_011514954.2 Intron XP_011513256.1
XM_017011369.1 Intron XP_016866858.1
XM_017011370.1 Intron XP_016866859.1
XM_017011371.1 Intron XP_016866860.1
XM_017011372.1 Intron XP_016866861.1
XM_017011373.1 Intron XP_016866862.1
XM_017011374.1 Intron XP_016866863.1

View Full Product Details