Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001098627.3 | Intron | NP_001092097.2 | |||
NM_001098629.2 | Intron | NP_001092099.1 | |||
NM_001098630.2 | Intron | NP_001092100.1 | |||
NM_001242452.2 | Intron | NP_001229381.1 | |||
NM_032643.4 | Intron | NP_116032.1 | |||
XM_005250317.3 | Intron | XP_005250374.1 | |||
XM_006715974.2 | Intron | XP_006716037.1 | |||
XM_011516158.2 | Intron | XP_011514460.1 | |||
XM_011516159.2 | Intron | XP_011514461.1 | |||
XM_011516160.1 | Intron | XP_011514462.1 | |||
XM_011516161.1 | Intron | XP_011514463.1 | |||
XM_011516162.1 | Intron | XP_011514464.1 | |||
XM_011516163.2 | Intron | XP_011514465.1 | |||
XM_011516164.1 | Intron | XP_011514466.1 |