Product Details

SNP ID
rs518844
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:182600284 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCCGGCTGGCTTCCTCACTGCC[A/G]TGGAGACTCAGGTGTGTGAGGGCTC
Phenotype
MIM: 602514
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RGS16 PubMed Links

Gene Details

Gene
RGS16
Gene Name
regulator of G-protein signaling 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002928.3 771 UTR 3 NP_002919.3

View Full Product Details