Product Details

SNP ID
rs208894
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.21:17442250 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAATCCAGCTGGGAGTCAGTGTATC[C/T]ATTTTATGTCATTTGCAGTAAATTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01549 PubMed Links

Gene Details

Gene
LINC01549
Gene Name
long intergenic non-protein coding RNA 1549
There are no transcripts associated with this gene.

View Full Product Details