Product Details
- SNP ID
-
rs686868
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:76819347 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CTGGGCAGACGGTGGGATCCGGGCA[A/G]GTGTGCGAAGGCTGTTCTGCTGCCG
- Phenotype
-
MIM: 600933
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
F2RL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111871548,rs113107082] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- F2RL1
- Gene Name
- F2R like trypsin receptor 1
View Full Product Details