Product Details
- SNP ID
-
hCV2383135
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:1207121 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAGGTGAAGGAGGTGCTGGACTCG[G/T]AGACGCTGTGCAGGAGGGCCGTCAA
- Phenotype
-
MIM: 602216
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
STK11
PubMed Links
Gene Details
- Gene
- STK11
- Gene Name
- serine/threonine kinase 11
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000455.4 |
1323 |
Nonsense Mutation |
GAG,TAG |
E70* |
NP_000446.1 |
View Full Product Details