Product Details

SNP ID
rs714601
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:56001548 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAAAAAATATATTGTGTGAGCTT[A/G]TATAGCTAAGAATTCCCAGCCCCCT
Phenotype
MIM: 600694
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FLJ31104 PubMed Links

Gene Details

Gene
FLJ31104
Gene Name
uncharacterized LOC441072
There are no transcripts associated with this gene.

Gene
IL6ST
Gene Name
interleukin 6 signal transducer
There are no transcripts associated with this gene.

View Full Product Details