Product Details

SNP ID
rs1074143
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:68093375 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTACAGGAGAGGAAATTGAGACTCA[A/G]ATTGACCTGCCCAAGGTCATTTAAC
Phenotype
MIM: 602698
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NFATC3 PubMed Links

Gene Details

Gene
NFATC3
Gene Name
nuclear factor of activated T-cells 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004555.3 Intron NP_004546.1
NM_173163.2 Intron NP_775186.1
NM_173165.2 Intron NP_775188.1

View Full Product Details