Product Details
- SNP ID
-
rs715112
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:35242731 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CCTCTAAGACCCCAGAGCTCCCTGA[A/C]TCTCTGCTCAAGCCCTCCTGTGACT
- Phenotype
-
MIM: 606061
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TBX20
PubMed Links
Gene Details
- Gene
- TBX20
- Gene Name
- T-box 20
View Full Product Details