Product Details

SNP ID
rs167060
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:30381909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGTTGTTTTTCTCATCATCATAC[G/T]TCTGTTATTTTCTTTCCTTGGCAGT
Phenotype
MIM: 609756
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CHRFAM7A PubMed Links

Gene Details

Gene
CHRFAM7A
Gene Name
CHRNA7 (exons 5-10) and FAM7A (exons A-E) fusion
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139320.1 Intron NP_647536.1
NM_148911.1 Intron NP_683709.1
Gene
LOC105370751
Gene Name
uncharacterized LOC105370751
There are no transcripts associated with this gene.

View Full Product Details