Product Details

SNP ID
rs606908
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54190589 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGCGGGGCCACAGGCCGCGAGG[C/G]TGCCGGGAGCCGATGACGCCCGAAC
Phenotype
MIM: 606048 MIM: 603631 MIM: 608754
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
MBOAT7 PubMed Links

Gene Details

Gene
MBOAT7
Gene Name
membrane bound O-acyltransferase domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146056.2 Intron NP_001139528.1
NM_001146082.2 Intron NP_001139554.1
NM_001146083.2 Intron NP_001139555.1
NM_024298.4 Intron NP_077274.3
XM_011527299.2 Intron XP_011525601.1
XM_011527300.2 Intron XP_011525602.1
XM_017027296.1 Intron XP_016882785.1
Gene
RPS9
Gene Name
ribosomal protein S9
There are no transcripts associated with this gene.

Gene
TSEN34
Gene Name
tRNA splicing endonuclease subunit 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077446.3 Intron NP_001070914.1
NM_001282332.1 Intron NP_001269261.1
NM_001282333.1 Intron NP_001269262.1
NM_024075.4 Intron NP_076980.2
XM_005278290.4 Intron XP_005278347.1
XM_011527294.2 Intron XP_011525596.1
XM_011527295.1 Intron XP_011525597.1

View Full Product Details