Product Details

SNP ID
rs231778
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:203869098 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGTGTGACCGCATTTAGTGGTGAT[A/G]AGTGTGAGCTTGCTTGGGGAGAGGG
Phenotype
MIM: 123890
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTLA4 PubMed Links

Gene Details

Gene
CTLA4
Gene Name
cytotoxic T-lymphocyte associated protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037631.2 Intron NP_001032720.1
NM_005214.4 Intron NP_005205.2

View Full Product Details