Product Details

SNP ID
rs680474
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:15515680 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCTGAAATCAGCATTTCGTGTTAC[C/T]GGTTATTAAGAGTATTGGGGAATAT
Phenotype
MIM: 605656
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXL7 PubMed Links

Gene Details

Gene
FBXL7
Gene Name
F-box and leucine rich repeat protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278317.1 Intron NP_001265246.1
NM_012304.4 Intron NP_036436.1
XM_011513998.1 Intron XP_011512300.1
XM_017009262.1 Intron XP_016864751.1

View Full Product Details