Product Details

SNP ID
rs442511
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6531085 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGACCCCGAAGCCCCGTGGCCT[C/G]CCGCGCCCCGCGCTCGCGCCTGCCG
Phenotype
MIM: 606182
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TNFSF9 PubMed Links

Gene Details

Gene
TNFSF9
Gene Name
tumor necrosis factor superfamily member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003811.3 87 Missense Mutation CCC,GCC P17A NP_003802.1

View Full Product Details