Product Details

SNP ID
rs11577661
Assay Type
Functionally tested
NCBI dbSNP Submissions
39
Location
Chr.1:54227863 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAAATGAGAATGCACACAGAAGC[C/T]CACTATAGAAAAGCAGACAGCCAAC
Phenotype
MIM: 611843 MIM: 607390
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPL37 PubMed Links

Gene Details

Gene
MRPL37
Gene Name
mitochondrial ribosomal protein L37
There are no transcripts associated with this gene.

Gene
SSBP3
Gene Name
single stranded DNA binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009955.3 Intron NP_001009955.1
NM_018070.4 Intron NP_060540.2
NM_145716.3 Intron NP_663768.1
XM_006710545.3 Intron XP_006710608.1
XM_017000897.1 Intron XP_016856386.1
XM_017000898.1 Intron XP_016856387.1
XM_017000899.1 Intron XP_016856388.1
Gene
SSBP3-AS1
Gene Name
SSBP3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details