Product Details

SNP ID
rs34410211
Assay Type
Functionally Tested
NCBI dbSNP Submissions
1
Location
Chr.6:31138111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGGATCCGTTCTAGGCGGTTCA[G/T]GGAGCCAGACTCCAGTTTCAGGCAG
Phenotype
MIM: 605310 MIM: 613525 MIM: 613526
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
CCHCR1 PubMed Links

Gene Details

Gene
CCHCR1
Gene Name
coiled-coil alpha-helical rod protein 1
There are no transcripts associated with this gene.

Gene
PSORS1C1
Gene Name
psoriasis susceptibility 1 candidate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014068.2 575 Intron NP_054787.2
Gene
PSORS1C2
Gene Name
psoriasis susceptibility 1 candidate 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014069.2 575 Missense Mutation CAT,CCT H84P NP_054788.2

View Full Product Details