Product Details

SNP ID
rs12482324
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:38230329 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATTGACAAGTTGTTTCTCTTCTT[C/T]GGGTAAATGTCTATTTAGATCCATT
Phenotype
MIM: 602106
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNJ15 PubMed Links

Gene Details

Gene
KCNJ15
Gene Name
potassium voltage-gated channel subfamily J member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276435.1 Intron NP_001263364.1
NM_001276436.1 Intron NP_001263365.1
NM_001276437.1 Intron NP_001263366.1
NM_001276438.1 Intron NP_001263367.1
NM_001276439.1 Intron NP_001263368.1
NM_002243.4 Intron NP_002234.2
NM_170736.2 Intron NP_733932.1
NM_170737.2 Intron NP_733933.1
XM_005260975.2 Intron XP_005261032.1
XM_006724002.2 Intron XP_006724065.1
XM_011529560.2 Intron XP_011527862.1
XM_011529561.2 Intron XP_011527863.1
XM_017028343.1 Intron XP_016883832.1
XM_017028344.1 Intron XP_016883833.1
XM_017028345.1 Intron XP_016883834.1

View Full Product Details