Product Details

SNP ID
rs11705205
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23262992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCAAAGGAGATGAGGGGAGCGTA[G/T]GGGCCGGGAAAGAGGCACAAGGAAG
Phenotype
MIM: 151410
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BCR PubMed Links

Gene Details

Gene
BCR
Gene Name
BCR, RhoGEF and GTPase activating protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004327.3 Intron NP_004318.3
NM_021574.2 Intron NP_067585.2
Gene
FBXW4P1
Gene Name
F-box and WD repeat domain containing 4 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details