Product Details

SNP ID
rs6905862
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31159825 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATACTCCCATCAGATTGGATGGAT[A/G]GTCGTGGTCCAGACCTTCATCTTCC
Phenotype
MIM: 605310 MIM: 164177 MIM: 600912
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCHCR1 PubMed Links

Gene Details

Gene
CCHCR1
Gene Name
coiled-coil alpha-helical rod protein 1
There are no transcripts associated with this gene.

Gene
POU5F1
Gene Name
POU class 5 homeobox 1
There are no transcripts associated with this gene.

Gene
TCF19
Gene Name
transcription factor 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077511.1 Intron NP_001070979.1
NM_001318908.1 Intron NP_001305837.1
NM_007109.2 Intron NP_009040.2

View Full Product Details