Product Details

SNP ID
rs2817213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:24495826 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGGCCCCTGGCCAGAATCATGG[A/G]ATCGGAAACGGAGAGAACTGTAGAA
Phenotype
MIM: 610045 MIM: 602515
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALDH5A1 PubMed Links

Gene Details

Gene
ALDH5A1
Gene Name
aldehyde dehydrogenase 5 family member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080.3 Intron NP_001071.1
NM_170740.1 Intron NP_733936.1
Gene
GPLD1
Gene Name
glycosylphosphatidylinositol specific phospholipase D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001503.3 Intron NP_001494.2
XM_011514509.2 Intron XP_011512811.1
XM_017010753.1 Intron XP_016866242.1
XM_017010754.1 Intron XP_016866243.1

View Full Product Details