Product Details

SNP ID
rs6039932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:10434849 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTATGTTCAAACATGGCTATTTT[C/T]ATTAAAATTTATCCGGCACTGTGAA
Phenotype
MIM: 604896 MIM: 615958
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MKKS PubMed Links

Gene Details

Gene
MKKS
Gene Name
McKusick-Kaufman syndrome
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018848.3 397 Intron NP_061336.1
NM_170784.2 397 Intron NP_740754.1
Gene
SLX4IP
Gene Name
SLX4 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009608.2 397 Intron NP_001009608.1
XM_017027647.1 397 Intron XP_016883136.1
XM_017027648.1 397 UTR 5 XP_016883137.1
XM_017027649.1 397 Intron XP_016883138.1
XM_017027650.1 397 UTR 5 XP_016883139.1
XM_017027651.1 397 Intron XP_016883140.1
XM_017027652.1 397 UTR 5 XP_016883141.1
XM_017027653.1 397 Intron XP_016883142.1

View Full Product Details