Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001304506.1 | 3054 | UTR 3 | NP_001291435.1 | ||
NM_019110.4 | 3054 | UTR 3 | NP_061983.2 | ||
XM_005249095.3 | 3054 | UTR 3 | XP_005249152.1 | ||
XM_005249097.3 | 3054 | UTR 3 | XP_005249154.1 | ||
XM_005249098.3 | 3054 | UTR 3 | XP_005249155.1 | ||
XM_005249100.4 | 3054 | UTR 3 | XP_005249157.2 | ||
XM_006715095.3 | 3054 | UTR 3 | XP_006715158.1 | ||
XM_011514588.2 | 3054 | UTR 3 | XP_011512890.1 | ||
XM_011514589.2 | 3054 | UTR 3 | XP_011512891.1 | ||
XM_017010844.1 | 3054 | UTR 3 | XP_016866333.1 |