Product Details
- SNP ID
-
rs6057649
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:31372655 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- ATGACTAAAGTTTCTTTTCATTGAT[A/C]GTAAGATTCATCCCACTTTCAGAAT
- Phenotype
-
MIM: 607650
MIM: 615997
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB118
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs73246868] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DEFB118
- Gene Name
- defensin beta 118
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_054112.2 |
|
Intron |
|
|
NP_473453.1 |
- Gene
- DEFB119
- Gene Name
- defensin beta 119
There are no transcripts associated with this gene.
View Full Product Details