Product Details

SNP ID
rs1011665
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27364519 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTCAGGCTTATTACAAGGATTAAA[C/T]GAGTTAACGTAAAGCACCTAGAACA
Phenotype
MIM: 603282
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF204P PubMed Links

Gene Details

Gene
ZNF204P
Gene Name
zinc finger protein 204, pseudogene
There are no transcripts associated with this gene.

View Full Product Details