Product Details

SNP ID
rs2073102
Assay Type
Functionally Tested
NCBI dbSNP Submissions
40
Location
Chr.1:19256836 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GATAGAGAACTGGTACATGATGCTC[C/G]TGATGGGCTGGGAGACAAAATGTTG
Phenotype
MIM: 608478 MIM: 616846
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
AKR7L PubMed Links
Additional Information
For this assay, SNP(s) [rs76434038] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
AKR7L
Gene Name
aldo-keto reductase family 7 like (gene/pseudogene)
There are no transcripts associated with this gene.

Gene
EMC1
Gene Name
ER membrane protein complex subunit 1
There are no transcripts associated with this gene.

Gene
MRTO4
Gene Name
MRT4 homolog, ribosome maturation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016183.3 Intron NP_057267.2
XM_006710675.3 Intron XP_006710738.1

View Full Product Details