Product Details

SNP ID
rs2268408
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41642981 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTGCCTTCCTCCCTCTCTAACAC[C/T]GCGGGGAGGAGGGGGAGCAGGGCAG
Phenotype
MIM: 604971
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MDFI PubMed Links

Gene Details

Gene
MDFI
Gene Name
MyoD family inhibitor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300804.1 Intron NP_001287733.1
NM_001300805.1 Intron NP_001287734.1
NM_001300806.1 Intron NP_001287735.1
NM_005586.3 Intron NP_005577.1
XM_005249117.3 Intron XP_005249174.1
XM_011514625.2 Intron XP_011512927.1
XM_011514626.2 Intron XP_011512928.1
XM_017010867.1 Intron XP_016866356.1

View Full Product Details