Product Details

SNP ID
rs2412059
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:2611011 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGTTGAGTTGTCTATTATTGTTCA[A/G]TTGTCTGTTTCTCTCTTCATTTCTG
Phenotype
MIM: 607542 MIM: 604115
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
KCNQ1 PubMed Links
Additional Information
For this assay, SNP(s) [rs117387809] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
KCNQ1
Gene Name
potassium voltage-gated channel subfamily Q member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000218.2 Intron NP_000209.2
NM_181798.1 Intron NP_861463.1
Gene
KCNQ1OT1
Gene Name
KCNQ1 opposite strand/antisense transcript 1 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details