Product Details

SNP ID
rs5910
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:44372421 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTGGCCGGTTCCGCTTGAAGAAGCC[A/G]ACCTGGGGGTACACGGGGGCCAAGG
Phenotype
MIM: 607759
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM171A2 PubMed Links

Gene Details

Gene
FAM171A2
Gene Name
family with sequence similarity 171 member A2
There are no transcripts associated with this gene.

Gene
ITGA2B
Gene Name
integrin subunit alpha 2b
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000419.4 2993 Silent Mutation GTC,GTT V1021V NP_000410.2
XM_011524749.1 2993 Silent Mutation GTC,GTT V987V XP_011523051.1
XM_011524750.1 2993 Silent Mutation GTC,GTT V982V XP_011523052.1

View Full Product Details