Product Details

SNP ID
rs12818212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:47756520 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAACAGCTCTACCAGGACTGACATG[G/T]GGACTCTCCTCTCAATTCTGATTGC
Phenotype
MIM: 606057 MIM: 612187
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RAPGEF3 PubMed Links

Gene Details

Gene
RAPGEF3
Gene Name
Rap guanine nucleotide exchange factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098531.2 Intron NP_001092001.1
NM_001098532.2 Intron NP_001092002.1
NM_006105.5 Intron NP_006096.2
XM_005268571.3 Intron XP_005268628.1
XM_011537752.1 Intron XP_011536054.1
XM_011537755.1 Intron XP_011536057.1
XM_011537758.2 Intron XP_011536060.1
XM_017018688.1 Intron XP_016874177.1
Gene
SLC48A1
Gene Name
solute carrier family 48 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017842.2 Intron NP_060312.2
XM_005269016.4 Intron XP_005269073.1
XM_011538558.2 Intron XP_011536860.1
XM_017019612.1 Intron XP_016875101.1
XM_017019613.1 Intron XP_016875102.1
XM_017019614.1 Intron XP_016875103.1
XM_017019615.1 Intron XP_016875104.1
XM_017019616.1 Intron XP_016875105.1
XM_017019617.1 Intron XP_016875106.1

View Full Product Details