Product Details

SNP ID
rs10403272
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19078507 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAGACTCTGTGCATATGGTCATCC[C/T]TGTGTGGGTCATTTGCAGTACCGAT
Phenotype
MIM: 610823
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC25A42 PubMed Links

Gene Details

Gene
SLC25A42
Gene Name
solute carrier family 25 member 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321544.1 Intron NP_001308473.1
NM_178526.4 Intron NP_848621.2
XM_005259861.3 Intron XP_005259918.1
XM_011527937.2 Intron XP_011526239.1
XM_017026670.1 Intron XP_016882159.1

View Full Product Details