Product Details

SNP ID
rs34606656
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:97010441 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACTGTAAGGATATTTTCTCTTAC[C/T]CCTTGGGATCCAGGCTCTGAGTCTC
Phenotype
MIM: 600028 MIM: 600030
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
DLX5 PubMed Links

Gene Details

Gene
DLX5
Gene Name
distal-less homeobox 5
There are no transcripts associated with this gene.

Gene
DLX6
Gene Name
distal-less homeobox 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005222.3 1276 UTR 3 NP_005213.3
Gene
DLX6-AS1
Gene Name
DLX6 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details