Product Details

SNP ID
rs758016
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3611188 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGCTCAGTGACTTTTGGTTCCCA[C/T]GACCTTGACAGAGACAGGGACCTCG
Phenotype
MIM: 188070
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CACTIN PubMed Links

Gene Details

Gene
CACTIN
Gene Name
cactin, spliceosome C complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080543.1 3065 UTR 3 NP_001074012.1
NM_021231.1 3065 Intron NP_067054.1
XM_011528160.2 3065 Intron XP_011526462.1
XM_011528161.2 3065 Intron XP_011526463.1
Gene
CACTIN-AS1
Gene Name
CACTIN antisense RNA 1
There are no transcripts associated with this gene.

Gene
TBXA2R
Gene Name
thromboxane A2 receptor
There are no transcripts associated with this gene.

View Full Product Details