Product Details

SNP ID
rs740404
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:2232050 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AGACACTCCCAGACTGAGGGGTGGT[A/G]TGTGGCGGGTGGCAGGGTGGCTGTG
Phenotype
MIM: 607375 MIM: 600796
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DOT1L PubMed Links

Gene Details

Gene
DOT1L
Gene Name
DOT1 like histone lysine methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032482.2 8995 UTR 3 NP_115871.1
XM_005259659.3 8995 UTR 3 XP_005259716.1
XM_005259660.3 8995 UTR 3 XP_005259717.1
XM_011528359.2 8995 UTR 3 XP_011526661.1
XM_011528360.1 8995 UTR 3 XP_011526662.1
XM_011528361.2 8995 UTR 3 XP_011526663.1
XM_017027366.1 8995 UTR 3 XP_016882855.1
Gene
MIR1227
Gene Name
microRNA 1227
There are no transcripts associated with this gene.

Gene
MIR6789
Gene Name
microRNA 6789
There are no transcripts associated with this gene.

Gene
PLEKHJ1
Gene Name
pleckstrin homology domain containing J1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300836.1 8995 Intron NP_001287765.1
NM_018049.2 8995 Intron NP_060519.1
XM_017026924.1 8995 Intron XP_016882413.1
XM_017026925.1 8995 Intron XP_016882414.1
XM_017026926.1 8995 Intron XP_016882415.1
XM_017026927.1 8995 Intron XP_016882416.1
XM_017026928.1 8995 Intron XP_016882417.1
Gene
SF3A2
Gene Name
splicing factor 3a subunit 2
There are no transcripts associated with this gene.

View Full Product Details